missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
155.00€ - 435.00€
Spécification
| Antigène | HLA-B7 |
|---|---|
| Clone | BB7.1 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
16370864
|
Affymetrix eBioscience
46-5917-41 |
25 Tests | |||||||
16380864
![]() |
Invitrogen™
46-5917-42 |
100 Tests |
435.00€
100 pièces |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Mouse Monoclonal Antibody
HLA-B7 is one of the polymorphic proteins of human MHC Class I found on all nucleated cells. This surface protein is a heterodimer consisting of the specific transmembrane HLA-B chain paired with Beta-2 microglobulin. Classical MHC Class I molecules play a central role in adaptive immunity by presenting to T cells antigenic peptides derived either from the cell itself (classical presentation) or extracellular antigens (cross-presentation). The ligands of MHC class I include T cell receptor (TCR) and a costimulatory molecule CD8. Hundreds of HLA-B alleles have been described. Typing for these polymorphisms is routinely done for transplantation. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
| HLA-B7 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P01889 | |
| HLA-B | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| BB7.1 | |
| Flow Cytometry | |
| PerCP-eFluor 710 | |
| Mouse | |
| Human | |
| 3106 | |
| Papain solubilized HLA-B7 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| HLA-B |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit