Learn More
HLA class I ABC Mouse anti-Human, CoraLite 488, Clone: 5C5B7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-66013-100UL
Description
The HLA class I ABC antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human HLA class I ABC. This antibody recognizes human, pig antigen. The HLA class I ABC antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Spécification
| HLA class I ABC | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3105 | |
| IgG2b |
| Immunofluorescence, Flow Cytometry | |
| 5C5B7 | |
| Mouse | |
| Primary | |
| Human, Pig |