missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DMA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11838-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognize the 33 kDa HLA-DMA and the 50-60 kDa dimer in Western blotting.
HLA-DMA belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DMA) and a beta chain (DMB), both anchored in the membrane. It is located in intracellular vesicles. DM plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail.Spécification
| HLA-DMA | |
| Polyclonal | |
| Unconjugated | |
| HLA-DMA | |
| D6S222E, DAMA 147C13.1 001, DAMC 226H3.5 001, DAQB 96B10.1 001, DASS 67H14.8 001, DMA, DMA protein, hCG_2001572, HLA DMA, HLA DMA protein, HLADM, HLA-DMA, RING6, XXbac BCX136A3.1 001, XXbac BPG181M17.3 001 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3108 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28067 | |
| HLA-DMA | |
| HLA-DMA Fusion Protein Ag2452 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu