missing translation for 'onlineSavingsMsg'
Learn More

HLM Antibody, Novus Biologicals™

Product Code. 18125472 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantité:
0.025 mg
0.1 mg
missing translation for 'unitSize'
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18125472 0.025 mg 25µg
18106601 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18125472 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP223625SS

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HLM Polyclonal specifically detects HLM in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigène HLM
Applications Western Blot
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot 1:500-1:1000
Formule PBS with 0.02% Sodium Azide
Alias de gène ORP4, ORP-4, OSBPL1, OSBPL4
Espèces hôtes Rabbit
Immunogène Reacts with residues 549-563 [EKGRWDEANTTEKQR] of the HLM protein.
Méthode de purification Affinity Purified
Quantité 0.025 mg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 23762
Espèces cibles Human
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.