missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGCL Recombinant Rabbit Monoclonal Antibody (22H11)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA550068
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HMGCL Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
The protein encoded by this gene belongs to the HMG-CoA lyase family. It is a mitochondrial enzyme that catalyzes the final step of leucine degradation and plays a key role in ketone body formation. Mutations in this gene are associated with HMG-CoA lyase deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification
| HMGCL | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35914 | |
| HMGCL | |
| A synthesized peptide derived from human HMGCL. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 22H11 | |
| Unconjugated | |
| HMGCL | |
| 3-hydroxy-3-methylglutarate-CoA lyase; 3-hydroxy-3-methylglutaryl-CoA lyase; 3-hydroxy-3-methylglutaryl-Coenzyme A lyase; 3-hydroxymethyl-3-methylglutaryl-CoA lyase; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase; AW476067; HL; Hmgcl; HMG-CoA lyase; hydroxymethylglutaricaciduria; hydroxymethylglutaryl-CoA lyase, mitochondrial; mitochondrial 3-hydroxy-3-methylglutaryl-CoA lyase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3155 | |
| -20°C or -80°C if preferred | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu