missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hormone-sensitive Lipase/HSL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Marque: Novus Biologicals NB110-37253SS
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot.
Spécification
| Hormone-sensitive Lipase/HSL | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| LIPE | |
| A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). [Swiss-Prot P15304] | |
| 0.025 mL | |
| Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| 3991 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu