missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ HPDL Antibody (SR2424), Novus Biologicals™

Product Code. 30041580 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30041580 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30041580 Supplier Novus Biologicals™ Supplier No. NBP327725

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

HPDL Monoclonal antibody specifically detects HPDL in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigène HPDL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SR2424
Conjugué Unconjugated
Dilution Western Blot 1:500-1:1000, Immunoprecipitation 1:20
Formule 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gène 4-hydroxyphenylpyruvate dioxygenase-like, 4-hydroxyphenylpyruvate dioxygenase-like protein, EC 1.13, GLOXD1,4-HPPD-L, glyoxalase domain containing 1, Glyoxalase domain-containing protein 1, MGC15668, RP4-534D1.1
Espèces hôtes Rabbit
Immunogène A synthetic peptide of human HPDL (Uniprot #: Q96IR7)
Méthode de purification Affinity purified
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 84842
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.