Learn More
Invitrogen™ HSP60 Monoclonal Antibody (2E1/53)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA3013
Description
MA3-013 detects HSP60 from human, monkey, mouse, and rat samples. This antibody displays no other protein reactivity. MA3-013 has been successfully used in Western blot, immunoprecipitation, immunohistochemistry, immunofluorescence, and ELISA procedures. By Western blot, this antibody detects a single 58 kDa protein representing recombinant human HSP60. The MA3-013 antigen is human placental HSP60. Epitope mapping studies using human HSP60 deletion mutants suggest that this antibody binds between amino acids 211-288. Reconstitute with PBS.
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Spécification
| HSP60 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P10809, P63038, P63039 | |
| HSPD1 | |
| Human placental HSP60. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgM |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2E1/53 | |
| Unconjugated | |
| HSPD1 | |
| 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) | |
| Mouse | |
| Purified | |
| RUO | |
| 15510, 3329, 63868 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.