Learn More
Human alpha 1-Microglobulin Antibody, R&D Systems™
Description
alpha 1-Microglobulin Monoclonal specifically detects alpha 1-Microglobulin in Human samples. It is validated for Western Blot, Immunocytochemistry.
Spécification
Spécification
| Antigène | alpha 1-Microglobulin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 784917 |
| Concentration | LYOPH |
| Conjugué | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Formule | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Numéro d’ordre du gène | P02760 |
| Alias de gène | A1M, alpha 1Microglobulin, alpha-1-microglobulin/bikunin precursor, AMBP, bikunin, EDC1, HCP, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITIL, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI |
| Afficher plus |