missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human alpha 2-Macroglobulin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems AF1938
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
alpha 2-Macroglobulin Polyclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.
Spécification
| alpha 2-Macroglobulin | |
| Polyclonal | |
| Unconjugated | |
| A2M, alpha 2Macroglobulin, Alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| A2M | |
| Human plasma-derived alpha 2 -Macroglobulin | |
| 100 μg | |
| Primary | |
| Detects human alpha 2 -Macroglobulin in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human alpha 2 -Macroglobulin-like 1 is observed. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu