missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-2/CD86 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Marque: R&D Systems MAB141-500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready, Knockout Validated.
Spécification
| B7-2/CD86 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Neutralization 0.5-2.5 ug/mL, CyTOF-ready, Knockout Validated | |
| P42081 | |
| CD86 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86 Ala23-His244 Accession # P42081 | |
| 500 μg | |
| Primary | |
| Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, recombinant mouse B7-2, recombinant rat B7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b, rhB7-H4, or rhB7-L2 is observed. | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, Neutralization, CyTOF, KnockDown | |
| 37301 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 942 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu