missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR10 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
310.00€ - 565.00€
Spécification
| Antigène | CCR10/GPR2 |
|---|---|
| Clone | 314305 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18711308
![]() |
R&D Systems
FAB3478G-025 |
25 Tests |
310.00€
25 tests |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18748947
![]() |
R&D Systems
FAB3478G-100 |
100 Tests |
565.00€
100 tests |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR10/GPR2 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
| CCR10 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CCR10. Stains human CCR10-transfected cells but not irrelevant transfectants. |
| 314305 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| Human | |
| P46092 | |
| 2826 | |
| Y3 rat myeloid cell line transfected with human CCR10 Gln8-Asn362 Accession # P46092 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title