Learn More
Human CCR10 PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 14 publications
Marque: R&D Systems FAB3478P-025
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Human samples. It is validated for Flow Cytometry.
Spécification
| CCR10/GPR2 | |
| Monoclonal | |
| R-PE | |
| P46092 | |
| CCR10 | |
| Y3 rat myeloid cell line transfected with human CCR10 Gln8-Asn362 Accession # P46092 | |
| 25 Tests | |
| 2826 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 314305 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detect human CCR10. Stains human CCR10-transfected cells but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction