missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD34 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB7227-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry.
Spécification
| CD34 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and Sodium Azide with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 947 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry | |
| QBEnd10 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
| Human endothelial vesicles | |
| 25 μg | |
| Primary | |
| Detects human CD34 in direct ELISAs | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu