missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD34 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB7227-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Flow Cytometry.
Spécification
| CD34 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and Sodium Azide with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 947 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry | |
| QBEnd10 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
| Human endothelial vesicles | |
| 100 μg | |
| Primary | |
| Detects human CD34 in direct ELISAs | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu