missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD52 (Research Grade Alemtuzumab Biosimilar) Biotinylated Antibody, R&D Systems™

Human Monoclonal Antibody
Marque: R&D Systems FAB9889B-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD52 Monoclonal specifically detects CD52 in Human samples. It is validated for Flow Cytometry.
Spécification
| CD52 | |
| Monoclonal | |
| Biotin | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human CD52 based on Alemtuzumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Alemtuzumab. This product is for research use only. | |
| Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu116 | |
| Flow Cytometry 10 μL/106 cells | |
| Cambridge pathology 1 antigen, CAMPATH-1, CAMPATH-1 antigen, CD52, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, HEL-S-171mP, Human epididymis-specific protein 5 | |
| Human CD52 | |
| 100 Tests | |
| 1043.0 | |
| Human | |
| Supernatant |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu