missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC10A/CD301 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB48881
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CLEC10A/CD301 Monoclonal specifically detects CLEC10A/CD301 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| CLEC10A/CD301 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q8IUN9 | |
| CLEC10A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC10A/CD301 Gln61-His316 Accession # Q8IUN9 | |
| 100 μg | |
| Primary | |
| Detects human CLEC10A/CD301 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ASGPR1 or recombinant mouse MGL1 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 744812 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD301, CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10462 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu