missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CMG-2/ANTXR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB29401-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CMG-2/ANTXR2 Monoclonal specifically detects CMG-2/ANTXR2 in Human samples. It is validated for ELISA.
Spécification
| CMG-2/ANTXR2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF, JHS | |
| Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2 Ser22-Arg200 Accession # P58335 | |
| 25 μg | |
| 118429 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 406510 | |
| ELISA | |
| P58335 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CMG-2/ANTXR2 in direct ELISAs. | |
| Human | |
| Supernatant |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu