missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C5/C5a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB20371-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Complement Component C5/C5a Monoclonal specifically detects Complement Component C5/C5a in Human samples. It is validated for ELISA.
Spécification
| Complement Component C5/C5a | |
| Monoclonal | |
| Unconjugated | |
| P01031 | |
| E. coli-derived recombinant human Complement Component C5a, Thr678-Arg751, Accession # P01031 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 295009 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 727 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu