missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL13/BLC/BCA-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Marque: R&D Systems AF801-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CXCL13/BLC/BCA-1 Polyclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Spécification
| CXCL13/BLC/BCA-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10563 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1-4 ug/mL | |
| Q53X90 | |
| CXCL13 | |
| E. coli-derived recombinant human CXCL13/BCA-1 Val23-Arg94 Accession # Q53X90 | |
| 25 μg | |
| Primary | |
| Detects CXCL13/BLC/BCA-1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu