missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL5/ENA-78 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems IC654R-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Spécification
| CXCL5/ENA-78 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P42830 | |
| CXCL5 | |
| E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 Accession # P42830 | |
| 100 μg | |
| Primary | |
| Detects human CXCL5/ENA-78 in ELISAs. Does not cross-react with recombinant human CXCL1, 2, or 3. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 33170 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6374 | |
| Human | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu