missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB5545
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Monoclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot.
Spécification
| Cytosolic Sulfotransferase 1E1/SULT1E1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P49888 | |
| SULT1E1 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 1E1/SULT1E1 Asn2-Ile294 Accession # P49888 | |
| 100 μg | |
| Primary | |
| Detects human Cytosolic Sulfotransferase 1E1/SULT1E1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SULT1A1, 1B1, 2A1, 2B1, or 4A1 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 726211 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STE, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4, SULT1E1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6783 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu