missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF5545
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Polyclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spécification
| Cytosolic Sulfotransferase 1E1/SULT1E1 | |
| Polyclonal | |
| Unconjugated | |
| P49888 | |
| SULT1E1 | |
| E. coli-derived recombinant human Sulfotransferase 1E1/SULT1E1 Asn2-Ile294 Accession # P49888 | |
| 100 μg | |
| Primary | |
| Detects human Sulfotransferase 1E1/SULT1E1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Sulfotransferase 1A1 is observed and less than 2% cross-reactivity with rhSulfotransferase 2A1 and rhSulfotransferase 4A1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STE, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4, SULT1E1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6783 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu