missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DPPIV/CD26 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems MAB11801-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human samples. It is validated for Immunocytochemistry.
Spécification
| DPPIV/CD26 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 | |
| Mouse myeloma cell line NS0-derived recombinant human DPPIV/CD26, Asp34-Pro766, Accession # Q53TN1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 222104 | |
| Immunocytochemistry 8-25 ug/mL | |
| Q53TN1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1803 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu