missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ECE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems MAB1784-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ECE-1 Monoclonal specifically detects ECE-1 in Human samples. It is validated for Western Blot.
Spécification
| ECE-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P42892 | |
| ECE1 | |
| Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 | |
| 25 μg | |
| Primary | |
| Detects human ECE-1 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) ECE-2 or rhMMEL-2. | |
| Human | |
| Purified |
| Western Blot | |
| 303908 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.71, ECE, ECE1, ECE-1, endothelin converting enzyme 1, endothelin-converting enzyme 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1889 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu