Learn More
Human EGFR (Research Grade Cetuximab Biosimilar) Biotinylated Antibody, R&D Systems™
Human Monoclonal Antibody
Marque: R&D Systems FAB9577B-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Spécification
| EGFR | |
| Monoclonal | |
| Biotin | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human EGFR based on Cetuximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Cetuximab. This product is for research use only. | |
| Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| Hu1 | |
| Flow Cytometry 10 uL/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Human EGFR | |
| 100 Tests | |
| 1956 | |
| Human | |
| Supernatant |