missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FABP2/I-FABP Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF3078
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Spécification
| FABP2/I-FABP | |
| Polyclonal | |
| Unconjugated | |
| P12104 | |
| FABP2 | |
| E. coli-derived recombinant human FABP2/I-FABP Ala2-Asp132 Accession # P12104 | |
| 100 μg | |
| Primary | |
| Detects human FABP2/I-FABP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant rat FABP2 is observed, 5% cross-reactivity with recombinant human (rh) FABP1, -4, -5, -7, and recombinant mouse FABP9 is observed, and less than 1% cross-reactivity with rhFABP3 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2169 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu