missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRL2/FcRH2, FCRL5/FcRH5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2048
This item is not returnable.
View return policy
Description
FCRL2/FcRH2 Monoclonal specifically detects FCRL2/FcRH2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| FCRL2/FcRH2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q96LA5 | |
| FCRL2 | |
| Mouse myeloma cell line NS0-derived recombinant human FCRL2/FcRH2 Glu15-Asp395 Accession # Q96LA5 | |
| 100 μg | |
| Primary | |
| Detects human FCRL2/FcRH2 and FCRL5/FcRH5 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human FCRL1, FCRL3, and FCRL4 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 296902 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AITRL, CD307b, CD307b antigen, Fc receptor homolog 2, Fc receptor-like 2, FcRH2, FCRH2SPAP1B, FcRL2, FcR-like protein 2, GITRL, IFGP family protein 4, IFGP4, Immunoglobulin receptor translocation-associated protein 4, immunoglobulin superfamily Fc receptor, gp42, IRTA4, IRTA4SPAP1C, SH2 domain containing phosphatase anchor protein 1, SH2 domain-containing phosphatase anchor protein 1Fc receptor-like protein 2, SPAP1, SPAP1A, SPAP1immune receptor translocation-associated protein 4, TNFSF18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 79368 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction