Learn More
Human FGF acidic/FGF1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB2321
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| FGF acidic/FGF1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2246 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| 939902 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| P05230 | |
| FGF1 | |
| E. coli-derived recombinant human FGF acidic/FGF1, Phe16-Asp155, Accession # P05230 | |
| 100 μg | |
| Primary | |
| Detects human FGF acidic/FGF1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Correction du contenu d'un produit
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu