Learn More
Human FGFR1 alpha (IIIc) Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems FAB11336R-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FGFR1 alpha Monoclonal antibody specifically detects FGFR1 alpha in Human samples. It is validated for Flow Cytometry
Spécification
| FGFR1 alpha | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2260 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1058809 | |
| Flow Cytometry | |
| P11362.3 | |
| Human embryonic kidney cell, HEK293-derived human FGFR1 alpha, Arg22-Glu376, Accession # P11362.3 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |