missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fibroblast Activation Protein alpha/FAP Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems FAB3715S-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Fibroblast Activation Protein alpha/FAP Monoclonal specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry.
Spécification
| Fibroblast Activation Protein alpha/FAP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q12884 | |
| FAP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha/FAP Leu26-Asp760 Accession # Q12884 | |
| RUO | |
| 2191 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 427819 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Fibroblast Activation Protein alpha/FAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human DPP6 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu