missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FIGLA Partial ORF (NP_001004311.1, 110 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marque:  Abnova™ H00344018-Q01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16152847

  • 508.00€ / 25µg
Expédition estimée: 05-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein that functions in postnatal oocyte-specific gene expression. The protein is a basic helix-loop-helix transcription factor that regulates multiple oocyte-specific genes, including genes involved in folliculogenesis and those that encode the zona pellucida. Mutations in this gene cause premature ovarian failure type 6. [provided by RefSeq]

Sequence: ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV
Spécification

Spécification

NP_001004311.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV
RUO
FIGLA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
344018
FIGLA (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
POF6/bHLHc8
FIGLA
Recombinant
wheat germ expression system
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions