missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human FOLR1 (Research Grade Mirvetuximab Biosimilar) Antibody, Novus Biologicals™

Human Monoclonal Antibody
Marque: Novus Biologicals™ MAB11490-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FOLR1 Monoclonal antibody specifically detects FOLR1 in Human samples. It is validated for ELISA
Spécification
| FOLR1 | |
| Monoclonal | |
| Unconjugated | |
| P15328 | |
| Human | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2348 | |
| Human | |
| Purified |
| ELISA | |
| Hu218 | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, Folbp1, FOLR, FR-alpha, KB cells FBP, MOv18, Ovarian tumor-associated antigen MOv18 | |
| Chinese hamster ovary cell line CHO-derived recombinant human FOLR1, Arg25-Met233, Accession # P15328 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu