missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FoxP3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems AF3240
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FoxP3 Polyclonal specifically detects FoxP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Spécification
| FoxP3 | |
| Polyclonal | |
| Unconjugated | |
| Q9BZS1 | |
| FOXP3 | |
| E. coli-derived recombinant human FoxP3 Gln105-Lys200 Accession # Q9BZS1 | |
| 100 μg | |
| Primary | |
| Detects human FoxP3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human FoxD3 is observed. | |
| Human | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50943 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu