missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFER/ALR Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB7940
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GFER/ALR Monoclonal specifically detects GFER/ALR in Human samples. It is validated for Western Blot, Immunohistochemistry.
Spécification
| GFER/ALR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P55789 | |
| GFER | |
| E. coli-derived recombinant human GFER/ALR Arg82-Asp205 Accession # P55789 | |
| 100 μg | |
| Primary | |
| Detects human GFER/ALR in ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 829812 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALR, ALRFAD-linked sulfhydryl oxidase ALR, Augmenter of liver regeneration, EC 1.8.3.2, ERV1, ERV1 homolog, ERV1hepatopoietin protein, erv1-like growth factor, growth factor, augmenter of liver regeneration, growth factor, erv1 (S. cerevisiae)-like (augmenter of liver regeneration), Hepatopoietin, hERV1, HERV1hepatic regenerative stimulation substance, HPO, HPO1, HPO2, HSS, truncated augmenter of liver regeneration | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2671 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu