missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF1439-SP
This item is not returnable.
View return policy
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Polyclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Human samples. It is validated for Western Blot.
Specifications
| GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64096 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NP_071422 | |
| GFRA4 | |
| Mouse myeloma cell line NS0-derived recombinant human GFR alpha-4/GDNF R alpha-4 Asn24-Ser245 (Ser140Cys) predicted Accession # NP_071422 | |
| 25 μg | |
| Primary | |
| Detects human GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) GFR alpha-1 is observed and less than 1% cross-reactivity with rhGFR alpha-2 and rhGFR alpha-3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction