Learn More
ACROBiosystems Human HBEGF Protein, His Tag
Brand: ACROBiosystems HBF-H52H3-1mg
Additional Details : Poids : 0.00010kg
Description
Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a ligand for the EGF receptor (EGFR) and a critical molecular component to a number of normal physiological processes including but not limited to tissue injury and wound healing, reproduction, angiogenesis and recently, adipogenesis. Misexpression of HB-EGF is linked to tumor formation and cancer including hepatocellular, pancreatic, gastric, breast, colon and melanoma, gliomas and glioblastomas.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.6 kDa.Specifications
11.6 kDa | |
Human HBEGF, His Tag (HBF-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 63 - Leu 148 (Accession # Q99075-1). | |
-20°C | |
HBEGF | |
Human | |
HEK293 | |
Cytokines&Growth; Factors |
1 mg | |
Asp 63 - Leu 148 | |
HBEGF, HB-EGF, Heparin-binding EGF-like growth factor, Diphtheria toxin receptor, DT-R, DTR, DTS, HEGFL | |
Unconjugated | |
His Tag | |
Powder |