missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human HBEGF Protein, His Tag
362.89€ - 2661.24€
Spécification
Poids moléculaire | 11.6 kDa |
---|---|
Source | Human HBEGF, His Tag (HBF-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 63 - Leu 148 (Accession # Q99075-1). |
Immunogène | Asp 63 - Leu 148 |
Conditions de stockage | -20°C |
Alias de gène | HBEGF, HB-EGF, Heparin-binding EGF-like growth factor, Diphtheria toxin receptor, DT-R, DTR, DTS, HEGFL |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17246220
|
ACROBiosystems
HBF-H52H3-100UG |
100 μg |
362.89€
100µg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17256220
|
ACROBiosystems
HBF-H52H3-1MG |
1 mg |
2661.24€
1mg |
Expédition estimée: 23-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Heparin-binding epidermal growth factor (EGF)-like growth factor (HBEGF) is a ligand for the EGF receptor (EGFR) and a critical molecular component to a number of normal physiological processes including but not limited to tissue injury and wound healing, reproduction, angiogenesis and recently, adipogenesis. Misexpression of HB-EGF is linked to tumor formation and cancer including hepatocellular, pancreatic, gastric, breast, colon and melanoma, gliomas and glioblastomas.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.6 kDa.Spécification
11.6 kDa | |
Asp 63 - Leu 148 | |
HBEGF, HB-EGF, Heparin-binding EGF-like growth factor, Diphtheria toxin receptor, DT-R, DTR, DTS, HEGFL | |
Unconjugated | |
His Tag | |
Powder |
Human HBEGF, His Tag (HBF-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 63 - Leu 148 (Accession # Q99075-1). | |
-20°C | |
HBEGF | |
Human | |
HEK293 | |
Cytokines&Growth; Factors |