Learn More
Abnova™ Human HCN4 Partial ORF (NP_005468.1, 1105 a.a. - 1203 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00010021-Q01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the hyperpolarization-activated cyclic nucleotide-gated potassium channels. The encoded protein shows slow kinetics of activation and inactivation, and is necessary for the cardiac pacemaking process. This channel may also mediate responses to sour stimuli. Mutations in this gene have been linked to sick sinus syndrome 2, also known as atrial fibrillation with bradyarrhythmia or familial sinus bradycardia. Two pseudogenes have been identified on chromosome 15. [provided by RefSeq]
Sequence: SPHSSGESMAAFPLFPRAGGGSGGSGSSGGLGPPGRPYGAIPGQHVTLPRKTSSGSLPPPLSLFGARATSSGGPPLTAGPQREPGARPEPVRSKLPSNLSpécification
NP_005468.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SPHSSGESMAAFPLFPRAGGGSGGSGSSGGLGPPGRPYGAIPGQHVTLPRKTSSGSLPPPLSLFGARATSSGGPPLTAGPQREPGARPEPVRSKLPSNL | |
RUO | |
HCN4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10021 | |
HCN4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HCN4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |