missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human HGFR/c-MET Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF276
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HGFR/c-MET Polyclonal specifically detects HGFR/c-MET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready, Knockout Validated.
Spécification
| HGFR/c-MET | |
| Polyclonal | |
| Unconjugated | |
| P08581 | |
| MET | |
| Mouse myeloma cell line NS0-derived recombinant human HGF R/c-MET Glu25-Thr932 Accession # P08581 | |
| 100 μg | |
| Primary | |
| Detects human HGF R/c-MET in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse HGF R is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL, CyTOF-ready, Knockout Validated | |
| AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4233 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu