Learn More
Human Hormone-sensitive Lipase/HSL Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marque: R&D Systems AF7104
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human samples. It is validated for Western Blot.
Spécification
| Hormone-sensitive Lipase/HSL | |
| Polyclonal | |
| Unconjugated | |
| Q05469 | |
| LIPE | |
| E. coli-derived recombinant human Hormone-sensitive Lipase/HSL Met302-Leu425 Accession # Q05469 | |
| 100 μg | |
| Primary | |
| Detects human Hormone-sensitive Lipase/HSL in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| hormone-sensitive lipase, Hormonesensitive Lipase, hormone-sensitive lipase testicular isoform, HSL, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive, LIPE | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3991 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |