missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17RA/IL-17R APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marque: R&D Systems FAB177A-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-17RA/IL-17R Monoclonal specifically detects IL-17RA/IL-17R in Human samples. It is validated for Flow Cytometry, Knockout Validated.
Spécification
| IL-17RA/IL-17R | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23765 | |
| Human | |
| Purified |
| Flow Cytometry, Immunoassay | |
| 133617 | |
| Flow Cytometry 10 uL/10^6 cells, Knockout Validated 10 uL/10^6 cells | |
| Q96F46 | |
| IL17RA | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 R Leu33-Trp320 Accession # Q96F46 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) IL-17 R, rmIL-17B R, or recombinant human IL-17B R is observed. In Western blots, no cross-reactivity with rmIL-17 R is observed | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu