missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-22 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Marque: R&D Systems MAB7821-500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-22 Monoclonal specifically detects IL-22 in Human samples. It is validated for Neutralization.
Spécification
| IL-22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50616 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 142928 | |
| Neutralization 1-5 ug/mL | |
| Q9GZX6 | |
| IL22 | |
| E. coli-derived recombinant human IL-22 Ala34-Ile179 Accession # Q9GZX6 | |
| 500 μg | |
| Primary | |
| Detects human IL-22 in direct ELISAs. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-22 and recombinant rat IL-22 and no cross-reactivity with recombinant human IL-10. | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu