missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha L/CD11a Antibody, R&D Systems™

Mouse Monoclonal Antibody
248.00€ - 549.00€
Spécification
| Antigène | Integrin alpha L/CD11a |
|---|---|
| Clone | 345908 |
| Dilution | Western Blot 2 μg/mL, Simple Western 50 μg/mL, Immunohistochemistry 5-15 μg/mL |
| Applications | Western Blot, Simple Western, Immunohistochemistry |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
18744364
|
R&D Systems
MAB35952-100 |
100 μg |
549.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18764394
![]() |
R&D Systems
MAB35952-SP |
25 μg |
248.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Integrin alpha L/CD11a Monoclonal antibody specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Western Blot, Simple Western, ImmunohistochemistrySpécification
| Integrin alpha L/CD11a | |
| Western Blot 2 μg/mL, Simple Western 50 μg/mL, Immunohistochemistry 5-15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD11 antigen-like family member A, CD11a, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), ITGAL, Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, Ly-15, Ly-21, lymphocyte function-associated antigen 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha L, Tyr26-Met1089 (Tyr660Ile), Accession # CAA68747 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 345908 | |
| Western Blot, Simple Western, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CAA68747 | |
| 3683 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit