Learn More
ACROBiosystems Human Integrin alpha X beta 2 (ITGAX&ITGB2) Heterodimer Protein, His Tag&Tag Free
Marque: ACROBiosystems IT2-H52W3-1mg
Informations supplémentaires : Poids : 0.00010kg
Description
The integrin αXβ2 takes a key role in host defense and functions as a receptor for the complement C3 cleavage product iC3b, fibrinogen, denatured proteins, and intercellular adhesion molecule (ICAM)-1. Integrin αXβ2 αXβ2 has a unique cellular distribution with expression on monocytes, macrophages, and the CD8− subsets of dendritic cells and involves in priming and regulation of the immune response.
Human ITGAX&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAX and ITGB2, has a calculated MW of 126.6 kDa (ITGAX) and 80.1 kDa (ITGB2). Subunit ITGAX is fused with an acidic tail at the C-terminus and followed by a polyhistidSpecifications
126.6 kDa, 80.1 kDa | |
Human ITGAX&ITGB2; Heterodimer Protein, His Tag&Tag; Free (IT2-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) (Accession # P20702-1 (ITGAX) & P05107-1 (ITGB2)). | |
-20°C | |
Integrin alpha x beta 2 | |
Human | |
HEK293 | |
Bio-Markers & CD Antigens |
1 mg | |
Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) | |
Integrin alpha x beta 2, ITGAX&ITGB2;, CD11C, CD18 | |
Unconjugated | |
His Tag | |
Powder |