missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human Integrin alpha X beta 2 (ITGAX&ITGB2) Heterodimer Protein, His Tag&Tag Free
495.95€ - 3870.90€
Spécification
Poids moléculaire | 126.6 kDa, 80.1 kDa |
---|---|
Source | Human ITGAX&ITGB2; Heterodimer Protein, His Tag&Tag; Free (IT2-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) (Accession # P20702-1 (ITGAX) & P05107-1 (ITGB2)). |
Immunogène | Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) |
Conditions de stockage | -20°C |
Alias de gène | Integrin alpha x beta 2, ITGAX&ITGB2;, CD11C, CD18 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17266280
|
ACROBiosystems
IT2-H52W3-1MG |
1 mg |
3870.90€
1mg |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17256280
|
ACROBiosystems
IT2-H52W3-100UG |
100 μg |
495.95€
100µg |
Expédition estimée: 28-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The integrin αXβ2 takes a key role in host defense and functions as a receptor for the complement C3 cleavage product iC3b, fibrinogen, denatured proteins, and intercellular adhesion molecule (ICAM)-1. Integrin αXβ2 αXβ2 has a unique cellular distribution with expression on monocytes, macrophages, and the CD8− subsets of dendritic cells and involves in priming and regulation of the immune response.
Human ITGAX&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAX and ITGB2, has a calculated MW of 126.6 kDa (ITGAX) and 80.1 kDa (ITGB2). Subunit ITGAX is fused with an acidic tail at the C-terminus and followed by a polyhistidSpécification
126.6 kDa, 80.1 kDa | |
Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) | |
Integrin alpha x beta 2, ITGAX&ITGB2;, CD11C, CD18 | |
Unconjugated | |
His Tag | |
Powder |
Human ITGAX&ITGB2; Heterodimer Protein, His Tag&Tag; Free (IT2-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) (Accession # P20702-1 (ITGAX) & P05107-1 (ITGB2)). | |
-20°C | |
Integrin alpha x beta 2 | |
Human | |
HEK293 | |
Bio-Markers & CD Antigens |