Learn More
ACROBiosystems Human Integrin alpha X beta 2 (ITGAX&ITGB2) Heterodimer Protein, His Tag&Tag Free
Marque:  ACROBiosystems IT2-H52W3-100ug
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17256280
  • 467.59€ / 100µg
Expédition estimée: 12-10-2022
pour voir le stock disponible 
Description
Description
The integrin αXβ2 takes a key role in host defense and functions as a receptor for the complement C3 cleavage product iC3b, fibrinogen, denatured proteins, and intercellular adhesion molecule (ICAM)-1. Integrin αXβ2 αXβ2 has a unique cellular distribution with expression on monocytes, macrophages, and the CD8− subsets of dendritic cells and involves in priming and regulation of the immune response.
Human ITGAX&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAX and ITGB2, has a calculated MW of 126.6 kDa (ITGAX) and 80.1 kDa (ITGB2). Subunit ITGAX is fused with an acidic tail at the C-terminus and followed by a polyhistid
Spécification
Spécification
Powder
100 μg
Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2)
Integrin alpha x beta 2, ITGAX&ITGB2;, CD11C, CD18
Unconjugated
HEK293
Human
126.6 kDa, 80.1 kDa
Human ITGAX&ITGB2; Heterodimer Protein, His Tag&Tag; Free (IT2-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) (Accession # P20702-1 (ITGAX) & P05107-1 (ITGB2)).
-20°C
Integrin alpha x beta 2
His Tag
Bio-Markers & CD Antigens