missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human L-Selectin/CD62L Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marque: R&D Systems MAB728-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| L-Selectin/CD62L | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1 | |
| Mouse myeloma cell line NS0-derived recombinant human L-selectin/CD62L, Trp39-Asn232, Accession # P14151 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 1009120 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P14151 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6402 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu