missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAG-3 PE-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Marque: R&D Systems FAB2319P
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LAG-3 Polyclonal specifically detects LAG-3 in Human samples. It is validated for Flow Cytometry.
Spécification
| LAG-3 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD223, CD223 antigen, FDC, LAG3, LAG-3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3902 | |
| Human | |
| IgG |
| Flow Cytometry | |
| R-PE | |
| P18627 | |
| LAG3 | |
| Mouse myeloma cell line NS0-derived recombinant human LAG-3 Leu23-Leu450 Accession # P18627 | |
| 100 Tests | |
| Primary | |
| Detects human LAG-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse LAG-3 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu