missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LDLR PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Marque: R&D Systems FAB2148P
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.
Spécification
| LDLR | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3949 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 472413 | |
| Flow Cytometry 10 uL/10^6 cells | |
| P01130 | |
| LDLR | |
| Chinese hamster ovary cell line CHO-derived recombinant human LDL R Ala22-Arg788 Accession # P01130 | |
| 100 Tests | |
| Primary | |
| Detects human LDL R in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) LDL R, recombinant human LRP-5, or rmLRP-6 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering